Thermo Scientific Chemicals

2-Carboxyphenyl phosphate, 98%, Thermo Scientific Chemicals

Catalog number: A14417.03
1 g, Each
Thermo Scientific Chemicals

2-Carboxyphenyl phosphate, 98%, Thermo Scientific Chemicals

Catalog number: A14417.03
1 g, Each
Quantity
Catalog number: A14417.03
also known as A14417-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6064-83-1
IUPAC Name
2-(phosphonooxy)benzoic acid
Molecular Formula
C7H7O6P
InChI Key
FFKUDWZICMJVPA-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CC=C1OP(O)(O)=O
Appearance (Color)
White to cream
Assay (HPLC)
≥97.5%
Melting Point (clear melt)
154.0-160.0?C
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

2-Carboxyphenyl phosphate is used as a substrate for the estimation of acid phosphatase. It is also used as a analgesic, anti-inflammatory agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Carboxyphenyl phosphate is used as a substrate for the estimation of acid phosphatase. It is also used as a analgesic, anti-inflammatory agent.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text