Thermo Scientific Chemicals

2,6-Dimethoxybenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: A14418.36
500 g, Each
Thermo Scientific Chemicals

2,6-Dimethoxybenzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: A14418.36
500 g, Each
Quantity
Catalog number: A14418.36
also known as A14418-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1466-76-8
IUPAC Name
2,6-dimethoxybenzoic acid
Molecular Formula
C9H10O4
InChI Key
MBIZFBDREVRUHY-UHFFFAOYSA-N
SMILES
COC1=CC=CC(OC)=C1C(O)=O
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥98.5%
Appearance (Color)
White to pale cream
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%

Description

It is used in the synthesis of indoles as well as coumarins related to Novobiocin, used as potent inhibitors of heat-shock protein 90 in the treatment of breast cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of indoles as well as coumarins related to Novobiocin, used as potent inhibitors of heat-shock protein 90 in the treatment of breast cancer.

Solubility
Insoluble in water. Solubility in methanol is almost transparent. Soluble in DMSO, Ethyl Acetate and acetone.

Notes
Store away from bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text