4-Methoxyphenoxyacetic acid, 98%, Thermo Scientific Chemicals
4-Methoxyphenoxyacetic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methoxyphenoxyacetic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number A14419.18
also known as A14419-18
Price (USD)/ Each
161.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
161.00
Add to cart
4-Methoxyphenoxyacetic acid, 98%, Thermo Scientific Chemicals
Catalog numberA14419.18
Price (USD)/ Each
161.00
-
Add to cart
Chemical Identifiers
CAS1877-75-4
IUPAC Name2-(4-methoxyphenoxy)acetic acid
Molecular FormulaC9H10O4
InChI KeyBHFSBJHPPFJCOS-UHFFFAOYSA-N
SMILESCOC1=CC=C(OCC(O)=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Melting Point (clear melt)107.0-114.0?C
Appearance (Color)Pale cream to cream to pale brown
Assay (Silylated GC)≥97.5%
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only