Thermo Scientific Chemicals

Triethyloxonium tetrafluoroborate, 95%, stab. with 3-5% diethyl ether, Thermo Scientific Chemicals

Catalog number: A14420.22
100 g, Each
Thermo Scientific Chemicals

Triethyloxonium tetrafluoroborate, 95%, stab. with 3-5% diethyl ether, Thermo Scientific Chemicals

Catalog number: A14420.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A14420.22
also known as A14420-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
368-39-8
IUPAC Name
tetrafluoroboranuide; triethyloxidanium
Molecular Formula
C6H15BF4O
InChI Key
IYDQMLLDOVRSJJ-UHFFFAOYSA-N
SMILES
F[B-](F)(F)F.CC[O+](CC)CC
Appearance (Color)
White to cream
Assay (Aqueous acid-base Titration)
>94.0 to <106.0%
Form
Crystals or Crystalline powder

Description

Triethyloxonium tetrafluoroborate is used as a powerful alkylating agent, especially for ethylation. It is used for the modification of carboxyl residues in proteins. It is involved in the preparation of μ -amino esters from lactams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyloxonium tetrafluoroborate is used as a powerful alkylating agent, especially for ethylation. It is used for the modification of carboxyl residues in proteins. It is involved in the preparation of µ -amino esters from lactams.

Solubility
Soluble in dichloromethane.

Notes
Moisture sensitive and hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text