Thermo Scientific Chemicals

Chromone-3-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: A14422.06
5 g, Each
Thermo Scientific Chemicals

Chromone-3-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: A14422.06
5 g, Each
Quantity
Catalog number: A14422.06
also known as A14422-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
39079-62-4
IUPAC Name
4-oxo-4H-chromene-3-carboxylate
Molecular Formula
C10H5O4
InChI Key
PCIITXGDSHXTSN-UHFFFAOYSA-M
SMILES
[O-]C(=O)C1=COC2=CC=CC=C2C1=O
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
198.0-205.0?C
Assay (HPLC)
≥97.5%
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Appearance (Color)
Pale cream to cream

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text