Linalool is used as a scent in 60-80% of perfumed hygiene products and cleaning agents including soaps, detergents, shampoos, and lotions. as a scent in 60-80% of perfumed hygiene products and cleaning agents including soaps, detergents, shampoos, and lotions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Linalool is used as a scent in 60-80% of perfumed hygiene products and cleaning agents including soaps, detergents, shampoos, and lotions. as a scent in 60-80% of perfumed hygiene products and cleaning agents including soaps, detergents, shampoos, and lotions.
Solubility
Soluble in water (1.589 g/L at 25°C).
Notes
Store at 4°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
A.T. Peana.; P.S. D’Aquila.; F. Panin.; G. Serra.; P. Pippia.; M.D.L. Moretti. Anti-inflammatory activity of linalool and linalyl acetate constituents of essential oils. Phytomedicine. 2002, 9 (8), 721-726.
C.F Bagamboula.; M Uyttendaele.; J Debevere. Inhibitory effect of thyme and basil essential oils, carvacrol, thymol, estragol, linalool and p-cymene towards Shigella sonnei and S. flexneri. Food Microbiology. 2004, 21 (1), 33-42.