Thermo Scientific Chemicals

1,1-Diphenylethylene, 98%, Thermo Scientific Chemicals

Catalog number: A14434.22
100 g, Each
Thermo Scientific Chemicals

1,1-Diphenylethylene, 98%, Thermo Scientific Chemicals

Catalog number: A14434.22
100 g, Each
Quantity
Catalog number: A14434.22
also known as A14434-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
530-48-3
IUPAC Name
(1-phenylethenyl)benzene
Molecular Formula
C14H12
InChI Key
ZMYIIHDQURVDRB-UHFFFAOYSA-N
SMILES
C=C(C1=CC=CC=C1)C1=CC=CC=C1
Form
Liquid
Refractive Index
1.6045-1.6095 @ 20?C
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥97.5%

Description

1,1-Diphenylethylene is used in the preparation of 2-chloro-1,1-diphenyl-ethene by reacting with benzeneseleninyl chloride and aluminum(III) chloride as reagents. It acts as an intermediate in organic synthesis as well as in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1-Diphenylethylene is used in the preparation of 2-chloro-1,1-diphenyl-ethene by reacting with benzeneseleninyl chloride and aluminum(III) chloride as reagents. It acts as an intermediate in organic synthesis as well as in pharmaceuticals.

Solubility
Miscible with methanol, chloroform and ether. Insoluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text