Thermo Scientific™

Copper(I) oxide, 97%, Thermo Scientific Chemicals

Catalog number: A14436.0E
2500 g, Each
Thermo Scientific™

Copper(I) oxide, 97%, Thermo Scientific Chemicals

Catalog number: A14436.0E
2500 g, Each
Quantity
Catalog number: A14436.0E
also known as A14436-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1317-39-1
Appearance (Color)
Red to orange-red
Assay (Redox Titration)
>96.0 to <104.0%
Elemental Analysis
Copper: Cu: ≥86.0%
Form
Powder

Description

It is used as a catalyst in decarboxylation reactions of arylacetic acids and of malonic acids under mild conditions. It is also used in batteries and electrodes, ceramic colorant, catalyst, purification of hydrogen, reagent in analytical chemistry, metallurgical and welding fluxes. It finds its application also as an antifouling agent for marine paints and in Benedict's Test. It is used to provide a source supplemental elemental copper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a catalyst in decarboxylation reactions of arylacetic acids and of malonic acids under mild conditions. It is also used in batteries and electrodes, ceramic colorant, catalyst, purification of hydrogen, reagent in analytical chemistry, metallurgical and welding fluxes. It finds its application also as an antifouling agent for marine paints and in Benedict′s Test. It is used to provide a source supplemental elemental copper.

Solubility
Insoluble in water. Dissolves in concentrated ammonia.

Notes
Air and moisture sensitive. Store away from air, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text