Thermo Scientific Chemicals

Aluminum chloride hexahydrate, 99%, Thermo Scientific Chemicals

Catalog number: A14437.0B
1000 g, Each
Thermo Scientific Chemicals

Aluminum chloride hexahydrate, 99%, Thermo Scientific Chemicals

Catalog number: A14437.0B
1000 g, Each
Quantity
Catalog number: A14437.0B
also known as A14437-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7784-13-6
IUPAC Name
aluminium(3+) hexahydrate trichloride
Molecular Formula
AlCl3H12O6
InChI Key
JGDITNMASUZKPW-UHFFFAOYSA-K
SMILES
O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]
Form
Crystals or Crystalline powder
Water Content (Karl Fischer Titration)
43.93 - 46.76% (5.8-6.5 waters, theory 44.77%)
Appearance (Color)
White
Assay (unspecified)
>98.5 to <101.5% (as hexahydrate)

Description

Pharmaceutical preparations of aluminum chloride hexahydrate are used to treat hyperhidrosis, a condition involving excessive sweating. It is also employed in deodorants, antiperspirant preparations, dyeing fabrics, refining crude oil, textile finishing, manufacturing parchment paper, preserving the wood and disinfecting stables. It also finds use as a reagent in organic synthesis. Aluminum chloride hexahydrate is also useful in water treatment process, medicine and cosmetic products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pharmaceutical preparations of aluminum chloride hexahydrate are used to treat hyperhidrosis, a condition involving excessive sweating. It is also employed in deodorants, antiperspirant preparations, dyeing fabrics, refining crude oil, textile finishing, manufacturing parchment paper, preserving the wood and disinfecting stables. It also finds use as a reagent in organic synthesis. Aluminum chloride hexahydrate is also useful in water treatment process, medicine and cosmetic products.

Solubility
Highly soluble in water. Soluble in alcohol, ether, glycerol, propylene glycol and acetonitrile.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text