2-Undecanol, 98+%, Thermo Scientific Chemicals
2-Undecanol, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Undecanol, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number A14441.18
also known as A14441-18
Price (USD)/ Each
231.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
231.00
Add to cart
2-Undecanol, 98+%, Thermo Scientific Chemicals
Catalog numberA14441.18
Price (USD)/ Each
231.00
-
Add to cart
Chemical Identifiers
CAS1653-30-1
IUPAC Nameundecan-2-ol
Molecular FormulaC11H24O
InChI KeyXMUJIPOFTAHSOK-UHFFFAOYNA-N
SMILESCCCCCCCCCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Refractive Index1.4350-1.4390 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
It was used to reduce tetrapropylammonium perruthenate in novel and efficient isomerization of allylic alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It was used to reduce tetrapropylammonium perruthenate in novel and efficient isomerization of allylic alcohols.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol and ethyl ether.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. István E. Markó, et al. Novel and Efficient Isomerization of Allylic Alcohols Promoted by a Tetrapropylammonium Perruthenate Catalyst.Angew. Chem. Int. Ed.,1999,38(13-14), 1960-1962.
  2. István E. Markó, et al. Efficient and Practical Catalytic Oxidation of Alcohols Using Molecular Oxygen.J. Org. Chem.,1988,66(22), 7576-7577.