Thermo Scientific Chemicals

Iodoethane, 98+%, stab. with copper, Thermo Scientific Chemicals

Catalog number: A14444.0E
2500 g, Each
Thermo Scientific Chemicals

Iodoethane, 98+%, stab. with copper, Thermo Scientific Chemicals

Catalog number: A14444.0E
2500 g, Each
Quantity
Catalog number: A14444.0E
also known as A14444-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-03-6
IUPAC Name
iodoethane
Molecular Formula
C2H5I
InChI Key
HVTICUPFWKNHNG-UHFFFAOYSA-N
SMILES
CCI
Appearance (Color)
Clear colorless to pale yellow
Identification (FTIR)
Conforms
Form
Liquid
Assay (GC)
≥98.0%
Refractive Index
1.5100-1.5140 @ 20?C

Description

Iodoethane is an excellent ethylating agent used in organic synthesis to introduce an ethyl group into a compound. It reacts with magnesium to form the Grignard reagent, ethylmagnesium iodide which is used in organic synthesis. It is involved in the preparation of 1-ethyl-3-nitro-2-phenyl-indole and also serves as hydrogen radical promoter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodoethane is an excellent ethylating agent used in organic synthesis to introduce an ethyl group into a compound. It reacts with magnesium to form the Grignard reagent, ethylmagnesium iodide which is used in organic synthesis. It is involved in the preparation of 1-ethyl-3-nitro-2-phenyl-indole and also serves as hydrogen radical promoter.

Solubility
Miscible with water.

Notes
Moisture and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong bases, strong oxidizing agents and magnesium.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text