Thermo Scientific Chemicals

Isophthalic acid, 99%, Thermo Scientific Chemicals

Catalog number: A14445.0B
1000 g, Each
Thermo Scientific Chemicals

Isophthalic acid, 99%, Thermo Scientific Chemicals

Catalog number: A14445.0B
1000 g, Each
Quantity
Catalog number: A14445.0B
also known as A14445-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
121-91-5
IUPAC Name
benzene-1,3-dicarboxylic acid
Molecular Formula
C8H6O4
InChI Key
QQVIHTHCMHWDBS-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC(=CC=C1)C(O)=O
Appearance (Color)
White
Form
Powder or crystals or crystalline powder
Assay from Supplier's CofA
≥98.5%

Description

Isophthalic acid is used as an intermediate for high performance unsaturated polyesters, resins for coatings, high solids paints, gel coats and modifier of polyethylene terephthalate for bottles. It acts as precursors for the fire-resistant material nomex as well as used in the preparation of high-performance polymer polybenzimidazole. It is also employed as an input for the production of insulation materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isophthalic acid is used as an intermediate for high performance unsaturated polyesters, resins for coatings, high solids paints, gel coats and modifier of polyethylene terephthalate for bottles. It acts as precursors for the fire-resistant material nomex as well as used in the preparation of high-performance polymer polybenzimidazole. It is also employed as an input for the production of insulation materials.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text