N-Acetyl-N'-phenylhydrazine, 98%, Thermo Scientific Chemicals
N-Acetyl-N'-phenylhydrazine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Acetyl-N'-phenylhydrazine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number A14446.22
also known as A14446-22
Price (USD)/ Each
96.50
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
96.50
Add to cart
N-Acetyl-N'-phenylhydrazine, 98%, Thermo Scientific Chemicals
Catalog numberA14446.22
Price (USD)/ Each
96.50
-
Add to cart
Chemical Identifiers
CAS114-83-0
IUPAC NameN'-phenylacetohydrazide
Molecular FormulaC8H10N2O
InChI KeyUICBCXONCUFSOI-UHFFFAOYSA-N
SMILESCC(=O)NNC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream
Assay (GC)≥97.5%
Melting Point (clear melt)127.0-133.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only