Thermo Scientific Chemicals

Phthalic acid, 99%, Thermo Scientific Chemicals

Catalog number: A14450.0I
5000 g, Each
Thermo Scientific Chemicals

Phthalic acid, 99%, Thermo Scientific Chemicals

Catalog number: A14450.0I
5000 g, Each
Quantity
Catalog number: A14450.0I
also known as A14450-0I
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
88-99-3
IUPAC Name
benzene-1,2-dicarboxylic acid
Molecular Formula
C8H6O4
InChI Key
XNGIFLGASWRNHJ-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CC=C1C(O)=O
Appearance (Color)
White
Assay (Silylated GC)
≥98.5%
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Identification (FTIR)
Conforms

Description

Phthalic acid is used as an active ingredient in polyesters and alkyd resins. It is also used as plasticizers for polyvinyl chloride and other polymers. It is also useful in the production of anthraquinone, phenolphthalein and phthalocyanine pigments. It serves as an organic reagent in the production of phthalates. It is used to prepare 1,3-cyclohexadiene by reduction using sodium amalgam.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phthalic acid is used as an active ingredient in polyesters and alkyd resins. It is also used as plasticizers for polyvinyl chloride and other polymers. It is also useful in the production of anthraquinone, phenolphthalein and phthalocyanine pigments. It serves as an organic reagent in the production of phthalates. It is used to prepare 1,3-cyclohexadiene by reduction using sodium amalgam.

Solubility
Soluble in water, alcohol and etherSoluble in alcohol. Slightly soluble in water, methanol and ether. Insoluble in chloroform.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text