Thermo Scientific Chemicals

2,2,2-Trifluoroacetamide, 97%, Thermo Scientific Chemicals

Catalog number: A14451.22
100 g, Each
Thermo Scientific Chemicals

2,2,2-Trifluoroacetamide, 97%, Thermo Scientific Chemicals

Catalog number: A14451.22
100 g, Each
Quantity
Catalog number: A14451.22
also known as A14451-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
354-38-1
Assay (HPLC)
≥96.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
68.0-77.0?C
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder

Description

2,2,2-Trifluoroacetamide used in a convenient alternative to the Gabriel synthesis of primary amines from halides by N-alkylation followed by cleavage of the readily-hydrolyzed trifluoroacetyl group and for improved N-alkylation under phase-transfer conditions allowing successive alkylation with different alkyl groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,2-Trifluoroacetamide used in a convenient alternative to the Gabriel synthesis of primary amines from halides by N-alkylation followed by cleavage of the readily-hydrolyzed trifluoroacetyl group and for improved N-alkylation under phase-transfer conditions allowing successive alkylation with different alkyl groups.

Solubility
Soluble in water (460 g/L at 20°C), alcohol, ether, chloroform (partially), and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids, strong bases and strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text