Thermo Scientific Chemicals

2-Fluoro-4-nitrotoluene, 98%, Thermo Scientific Chemicals

Catalog number: A14464.22
100 g, Each
Thermo Scientific Chemicals

2-Fluoro-4-nitrotoluene, 98%, Thermo Scientific Chemicals

Catalog number: A14464.22
100 g, Each
Quantity
Catalog number: A14464.22
also known as A14464-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1427-07-2
IUPAC Name
2-fluoro-1-methyl-4-nitrobenzene
Molecular Formula
C7H6FNO2
InChI Key
WIQISTBTOQNVCE-UHFFFAOYSA-N
SMILES
CC1=CC=C(C=C1F)[N+]([O-])=O
Melting Point (clear melt)
32.0-38.0?C
Appearance (Color)
Pale yellow to yellow to pale brown
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder or fused solid

Description

It finds its use in the synthesis of 2-fluoro-4-nitrobenzoic acid and also was used in a study of aquatic toxicity against Poecilia reticulata and Tetrahymena pyriformis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its use in the synthesis of 2-fluoro-4-nitrobenzoic acid and also was used in a study of aquatic toxicity against Poecilia reticulata and Tetrahymena pyriformis.

Solubility
Insoluble in water. Solubility in methanol gives very faint turbidity.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text