Thermo Scientific Chemicals

Silicon carbide powder, coarse, 46 grit, Thermo Scientific Chemicals

Catalog number: A14470.36
500 g, Each
Thermo Scientific Chemicals

Silicon carbide powder, coarse, 46 grit, Thermo Scientific Chemicals

Catalog number: A14470.36
500 g, Each
Quantity
Catalog number: A14470.36
also known as A14470-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
409-21-2
IUPAC Name
methanidylidynesilylium
Molecular Formula
CSi
InChI Key
HBMJWWWQQXIZIP-UHFFFAOYSA-N
SMILES
[C-]#[Si+]
Comment
Hardness: 9 moh / 2500 knoop diamond
Assay from Supplier's CofA
≥98.0% SiC
Impurity content
Iron(III) oxide: ≤0.2%
Particle Size
Average grain size 300-425µm by FEPA-Standard 42-1:2006
Comment
Accepted on supplier's Certificate of Analysis

Description

Silicon carbide is used in abrasives, in polishing and grinding. It is widely used in applications calling for high endurance, such as automobile brakes, car clutches and ceramic plates in bulletproof vests. Electronic applications of silicon carbide are as light emitting diodes and sensors. It is utilized for the production of fertile, transgenic maize plants. It compositely plays an important role in the crack-healing behavior.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Silicon carbide is used in abrasives, in polishing and grinding. It is widely used in applications calling for high endurance, such as automobile brakes, car clutches and ceramic plates in bulletproof vests. Electronic applications of silicon carbide are as light emitting diodes and sensors. It is utilized for the production of fertile, transgenic maize plants. It compositely plays an important role in the crack-healing behavior.

Solubility
Soluble in molten sodium hydroxide, potassium hydroxide and in molten iron.

Notes
Keep it away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text