Thermo Scientific Chemicals

Benzoylacetonitrile, 98+%, Thermo Scientific Chemicals

Catalog number: A14473.14
25 g, Each
Thermo Scientific Chemicals

Benzoylacetonitrile, 98+%, Thermo Scientific Chemicals

Catalog number: A14473.14
25 g, Each
Quantity
Catalog number: A14473.14
also known as A14473-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
614-16-4
IUPAC Name
3-oxo-3-phenylpropanenitrile
Molecular Formula
C9H7NO
InChI Key
ZJRCIQAMTAINCB-UHFFFAOYSA-N
SMILES
O=C(CC#N)C1=CC=CC=C1
Appearance (Color)
White to cream to pale brown
Assay (GC)
≥98.0%
Melting Point (clear melt)
78.0-84.0?C
Form
Crystals or powder or crystalline powder

Description

Benzoylacetonitrile is used to produce 2-benzoyl-3-furan-2-yl-acrylonitrile. It is also used in the synthesis of substituted naphtho[1,8-bc]pyrans and as building block in the preparation of 4H-pyrans, 2-pyridones, furans and carbocyclics. It is an active methylene compound, useful in heterocyclic synthesis, e.g. polyfunctional pyridines and pyrimidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzoylacetonitrile is used to produce 2-benzoyl-3-furan-2-yl-acrylonitrile. It is also used in the synthesis of substituted naphtho[1,8-bc]pyrans and as building block in the preparation of 4H-pyrans, 2-pyridones, furans and carbocyclics. It is an active methylene compound, useful in heterocyclic synthesis, e.g. polyfunctional pyridines and pyrimidines.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text