Thermo Scientific Chemicals

Hydrogen bromide, 33% w/w (45% w/v) soln. in acetic acid, Thermo Scientific Chemicals

Catalog number: A14475.AE
100 mL, Each
Thermo Scientific Chemicals

Hydrogen bromide, 33% w/w (45% w/v) soln. in acetic acid, Thermo Scientific Chemicals

Catalog number: A14475.AE
100 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A14475.AE
also known as A14475-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
37348-16-6
Form
fuming liquid
Appearance (Color)
Pale to dark yellow
Assay (Titration ex Bromide)
31 - 34.5% w/w HBr

Description

Hydrobromic acid is an indispensable raw material for organic intermediates, inorganic pharmaceuticals, photosensitive materials, dyes and medicines. It is also used in the production of bromine compounds. It is used as an important agent in the industrial processes of isomerization, polymerization, hydration and dehydration and esterification reactions. It plays an essential role for the extraction of mercury from the cinnebar ore.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text