Thermo Scientific Chemicals

Cyclobutanecarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: A14480.22
100 g, Each
Thermo Scientific Chemicals

Cyclobutanecarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: A14480.22
100 g, Each
Quantity
Catalog number: A14480.22
also known as A14480-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3721-95-7
IUPAC Name
cyclobutanecarboxylic acid
Molecular Formula
C5H8O2
InChI Key
TXWOGHSRPAYOML-UHFFFAOYSA-N
SMILES
OC(=O)C1CCC1
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.4415-1.4455 @ 20?C
Form
Liquid
Assay (Silylated GC)
≥98.0%
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%

Description

Cyclobutanecarboxylic acid is used in water treatment, row materials for sodium fluosilicate potassium fluosilicateammonium fluosilicatecopper fluosilicatebarium fluosilicate and other fluosilicates and silicic tetrafluoride. It is also used in refining lead of electrolysis, as mordant and in the treatment of metal surface.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclobutanecarboxylic acid is used in water treatment ,row materials for sodium fluosilicate potassium fluosilicateammonium fluosilicatecopper fluosilicatebarium fluosilicate and other fluosilicates and silicic tetrafluoride. It is also used in metal platingtimber antisepsisdisinfectant(1-2% of solution) of beer and brewing industry, refining lead of electrolysis, as mordant and in the treatment of metal surface.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text