Magnesium sulfate heptahydrate, 99+%, Thermo Scientific Chemicals
Magnesium sulfate heptahydrate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Magnesium sulfate heptahydrate, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
5000 g
Catalog number A14491.0B
also known as A14491-0B
Price (USD)/ Each
43.65
Online exclusive
48.40 
Save 4.75 (10%)
-
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (USD)/ Each
43.65
Online exclusive
48.40 
Save 4.75 (10%)
Add to cart
Magnesium sulfate heptahydrate, 99+%, Thermo Scientific Chemicals
Catalog numberA14491.0B
Price (USD)/ Each
43.65
Online exclusive
48.40 
Save 4.75 (10%)
-
Add to cart
Chemical Identifiers
CAS10034-99-8
IUPAC Namemagnesium(2+) heptahydrate sulfate
Molecular FormulaH14MgO11S
InChI KeyWRUGWIBCXHJTDG-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Complexometric Titration≥99.0%
FormCrystals or powder or crystalline powder or fused solid
Identification (FTIR)Conforms
Magnesium sulfate, produced by appropriate processes, finds use in several pharmaceutical preparations of magnesium usually known as epsom salt. It is used as a micronutrient in agriculture, for brewing salt in the production of certain alcoholic products, as an electrolyte to prepare metal salts, as a coagulant and as an ingredient in bath salts for use in floatation therapy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium sulfate, produced by appropriate processes, finds use in several pharmaceutical preparations of magnesium usually known as epsom salt. It is used as a micronutrient in agriculture, for brewing salt in the production of certain alcoholic products, as an electrolyte to prepare metal salts, as a coagulant and as an ingredient in bath salts for use in floatation therapy.

Solubility
Soluble in in water and acid. Slightly soluble in alcohol and glycerol.

Notes
Incompatible with strong oxidizers.
RUO – Research Use Only

General References:

  1. Abdulatif, M.; Amin, S. M. M.; Aboul-Ela, A.; Samuel, E. W. M.; Abdel-Hakim, S. M. A. Intra-articular versus intravenous magnesium-sulfate as adjuvant to femoral nerve block in arthroscopic knee surgery under general anesthesia: Randomized controlled trial. Egypt. J. Anaesth. 2015, 31, (3), 239-246.
  2. Puay, N. Q.; Qiu, G.; Ting, Y. P. Effect of Zinc oxide nanoparticles on biological wastewater treatment in a sequencing batch reactor. J. Clean. Prod. 2015, 88, 139-145.