Thermo Scientific Chemicals

Flavin adenine dinucleotide disodium salt hydrate, 94% (dry wt.), water <10%, Thermo Scientific Chemicals

Catalog number: A14495.MC
100 mg, Each
Thermo Scientific Chemicals

Flavin adenine dinucleotide disodium salt hydrate, 94% (dry wt.), water <10%, Thermo Scientific Chemicals

Catalog number: A14495.MC
100 mg, Each
Quantity
Catalog number: A14495.MC
also known as A14495-MC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1891059-93-0
Appearance (Color)
Yellow-orange
Water Content (Karl Fischer Titration)
<10.0%
pH
5.5 - 6.5
Form
Powder
Identification (FTIR)
Conforms

Description

Flavin adenine dinucleotide (FAD) is used as a redox cofactor (electron carrier) by flavoproteins including succinate dehydrogenase (complex), α-ketoglutarate dehydrogenase, apoptosis-inducing factor 2 (AIF-M2, AMID), folate/FAD-dependent tRNA methyltransferases, and N-hydroxylating flavoprotein monooxygenases. FAD is a component of the pyruvate dehydrogenase complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Flavin adenine dinucleotide (FAD) is used as a redox cofactor (electron carrier) by flavoproteins including succinate dehydrogenase (complex), α-ketoglutarate dehydrogenase, apoptosis-inducing factor 2 (AIF-M2, AMID), folate/FAD-dependent tRNA methyltransferases, and N-hydroxylating flavoprotein monooxygenases. FAD is a component of the pyruvate dehydrogenase complex.

Solubility
Soluble in water (50 mg/ml).

Notes
Store at 4°C. Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text