Thermo Scientific Chemicals

Diethyl oxalate, 99%, Thermo Scientific Chemicals

Catalog number: A14498.36
500 g, Each
Thermo Scientific Chemicals

Diethyl oxalate, 99%, Thermo Scientific Chemicals

Catalog number: A14498.36
500 g, Each
Quantity
Catalog number: A14498.36
also known as A14498-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
95-92-1
IUPAC Name
diethyl oxalate
Molecular Formula
C6H10O4
InChI Key
WYACBZDAHNBPPB-UHFFFAOYSA-N
SMILES
CCOC(=O)C(=O)OCC
Appearance (Color)
Clear colorless
Comment
Specification differs for U.S. and non-U.S. material where indicated
Refractive Index
1.4085-1.4120 @ 20°C [non-U.S. specification]
Assay from Suppliers CofA
≥98.5% (U.S. specification)
Assay (GC)
≥98.5%

Description

Diethyl oxalate is used to prepare active pharmaceutical ingredients (API), plastics and dyestuff intermediates. It is also used as a solvent for cellulose esters, ethers, resins, perfumes and lacquers for electronics. It is involved in the transesterification reaction with phenol to get dipheny oxalate. It is also involved in the Claisen condensation ketosteroids to prepare glyoxalyl derivatives. Further, it is used to prepare sym-1,4-diphenyl-1,4-dihydro-1,2,4,5-polytetrazine. In addition to this, it is utilized in the microemulsion synthesis of zinc oxide nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl oxalate is used to prepare active pharmaceutical ingredients (API), plastics and dyestuff intermediates. It is also used as a solvent for cellulose esters, ethers, resins, perfumes and lacquers for electronics. It is involved in the transesterification reaction with phenol to get dipheny oxalate. It is also involved in the Claisen condensation ketosteroids to prepare glyoxalyl derivatives. Further, it is used to prepare sym-1,4-diphenyl-1,4-dihydro-1,2,4,5-polytetrazine. In addition to this, it is utilized in the microemulsion synthesis of zinc oxide nanoparticles.

Solubility
Miscible with alcohols, ether and other common organic solvents.

Notes
Incompatible with strong oxidizing agents, reducing agents, acids and bases. Moisture sensitive.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text