Thermo Scientific Chemicals

Vitamin E acetate, 97%, Thermo Scientific Chemicals

Catalog number: A14505.36
500 g, Each
Thermo Scientific Chemicals

Vitamin E acetate, 97%, Thermo Scientific Chemicals

Catalog number: A14505.36
500 g, Each
Quantity
Catalog number: A14505.36
also known as A14505-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7695-91-2
IUPAC Name
(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-yl acetate
Molecular Formula
C31H52O3
InChI Key
ZAKOWWREFLAJOT-CEFNRUSXSA-N
SMILES
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(OC(C)=O)=C(C)C(C)=C2O1
Appearance (Color)
Clear, colorless to pale yellow to pale green
Assay from Supplier's CofA
≥96.0%
Form
Viscous liquid

Description

Vitamin E acetate is used as a fortifying agent and dietary supplement in food. It is used in dermatological products such as skin creams. It is an alternative to tocopherol and provides protection against the sun’s ultraviolet rays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vitamin E acetate is used as a fortifying agent and dietary supplement in food. It is used in dermatological products such as skin creams. It is an alternative to tocopherol and provides protection against the sun’s ultraviolet rays.

Solubility
Immiscible with water.

Notes
Store in a cool place. Air and light sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text