Thermo Scientific Chemicals

4-Chlorophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: A14518.06
5 g, Each
Thermo Scientific Chemicals

4-Chlorophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: A14518.06
5 g, Each
Quantity
Catalog number: A14518.06
also known as A14518-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1073-70-7
IUPAC Name
hydrogen (4-chlorophenyl)hydrazine chloride
Molecular Formula
C6H8Cl2N2
InChI Key
YQVZREHUWCCHHX-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NNC1=CC=C(Cl)C=C1
Form
Powder or lumps
Appearance (Color)
White to cream to yellow to pink to pale brown or purple
Assay (HPLC)
≥96.0%
Water Content (Karl Fischer Titration)
≤3%

Description

4-Chlorophenylhydrazine hydrochloride is used as a pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorophenylhydrazine hydrochloride is used as a pharmaceutical intermediates.

Solubility
Soluble in hot water, methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text