2-Bromobenzoyl chloride, 98%, Thermo Scientific Chemicals
2-Bromobenzoyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromobenzoyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A14520.09
also known as A14520-09
Price (USD)/ Each
60.10
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
60.10
Add to cart
2-Bromobenzoyl chloride, 98%, Thermo Scientific Chemicals
Catalog numberA14520.09
Price (USD)/ Each
60.10
-
Add to cart
Chemical Identifiers
CAS7154-66-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to orange
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.5945-1.5985 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only