Thermo Scientific Chemicals

2-Hydroxypyridine, 98%, Thermo Scientific Chemicals

Catalog number: A14522.14
25 g, Each
Thermo Scientific Chemicals

2-Hydroxypyridine, 98%, Thermo Scientific Chemicals

Catalog number: A14522.14
25 g, Each
Quantity
Catalog number: A14522.14
also known as A14522-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
142-08-5
Identification (FTIR)
Conforms
Form
Crystalline powder or crystals
Comment
Melting point is sensitive to water content
Melting Point (clear melt)
102-111?C
Appearance (Color)
White to cream to yellw to orange to pale brown

Description

2-Hydroxypyridine is a bifunctional catalyst for a wide variety of acylation reactions, catalyst for generating β-oxopropyl carbonates from cyclic carbonates and alcohols and in the aminolysis of a polyglutamate. It is used in peptide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxypyridine is a bifunctional catalyst for a wide variety of acylation reactions, catalyst for generating β-oxopropyl carbonates from cyclic carbonates and alcohols and in the aminolysis of a polyglutamate. It is used in peptide synthesis.

Solubility
Soluble in water 450 g/L @ 20°C and ethanol, slightly soluble in benzene and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text