Thermo Scientific Chemicals

4-Chlorobenzonitrile, 99%, Thermo Scientific Chemicals

Catalog number: A14524.18
50 g, Each
Thermo Scientific Chemicals

4-Chlorobenzonitrile, 99%, Thermo Scientific Chemicals

Catalog number: A14524.18
50 g, Each
Quantity
Catalog number: A14524.18
also known as A14524-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
623-03-0
IUPAC Name
4-chlorobenzonitrile
Molecular Formula
C7H4ClN
InChI Key
GJNGXPDXRVXSEH-UHFFFAOYSA-N
SMILES
ClC1=CC=C(C=C1)C#N
Assay (GC)
≥98.5%
Melting Point (clear melt)
89.0-95.0?C
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder or flakes or granules

Description

4-Chlorobenzonitrile is used to prepare a red pigment for plastics. It is also used in the preparation of amides in organic synthesis. Further, it is used as an intermediate of dye and active pharmaceutical ingredients.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorobenzonitrile is used to prepare a red pigment for plastics. It is also used in the preparation of amides in organic synthesis. Further, it is used as an intermediate of dye and active pharmaceutical ingredients.

Solubility
Soluble in alcohol and ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text