Informational:Interested in cGMP-banked Expi293F cells? Click here for product information.
Informational:Would you like training with your Expi293 system kit? Discover how »
Gibco™

ExpiFectamine™ 293 Transfection Kit

Catalog number: A14525
Have Questions?
Gibco™

ExpiFectamine™ 293 Transfection Kit

Catalog number: A14525
Have Questions?
Catalog Number
A14525
Unit Size
For 10 L of culture
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
For 50 L of culture
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
For 1 L of culture
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A14525For 10 L of culture
-
A14526For 50 L of culture
-
A14524For 1 L of culture
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The ExpiFectamine™ 293 Transfection Kit is a core component of the Expi293™ Expression System. It is designed for transient transfection of high density cultures of HEK (human embryonic kidney) 293 cells. The high-efficiency, cationic, lipid-based transfection reagent and transfection enhancers are designed to power the highest possible level of protein expression from Expi293F™ cells cultured in Expi293™ Expression Medium

Designed specifically for transfection of high density suspension cell culture, with matching transfection enhancers that boost transfection performance and protein expression
Achieves protein yields 2- to 10-fold higher than other transfection reagents used on high density 293 cell cultures
Employs the same transient expression protocols typically used in current low-density 293 suspension culture systems to easily switch from low density systems to the high yield, high density Expi293™ Expression System
Provides robust and reproducible transfection results, giving users greater confidence in their results
Scale transfections for culture volumes of less than 1 mL to greater than 10 liters, while maintaining equivalent volumetric protein yields

The ExpiFectamine™ 293 Transfection Kit is specifically designed to work with Expi293F™ cells in the Expi293™ Expression System. The transfection kit consists of the cationic lipid-based ExpiFectamine™ 293 Reagent and ExpiFectamine™ 293 Transfection Enhancers 1 and 2. The enhancers are added 16 to 18 hours post-transfection to boost transfection, cell viability, and protein expression. The protocol for use of ExpiFectamine™ 293 Transfection components is very similar to transfection protocols for other transient expression reagents, such as 293fectin™ and FreeStyle™ MAX.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Line
Expi293F™
Classification
Animal Origin-Free, Chemically Defined, Protein-Free, Serum-Free
Cell Type
Established Cell Lines
Sample Type
Plasmid DNA
Transfection Technique
Lipid-based Transfection
For Use With (Application)
Transfection
Product Line
ExpiFectamine™
Product Type
Transfection Kit
Purity
Cell Culture Grade
Quantity
For 10 L of culture
Serum Compatible
No

Contents & Storage

27 mL ExpiFectamine™ 293 Reagent
50 mL ExpiFectamine™ 293 Transfection Enhancer 1
500 mL ExpiFectamine™ 293 Transfection Enhancer 2

Sufficient for transfection of 10 L of culture

Store at +2°C to +8°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text