Expi293F™ Cells
Interested in cGMP-banked Expi293F cells? Click HERE for product information.
Expi293F™ Cells
Gibco™
Expi293F™ Cells
Expi293F human cells are derived from the 293 cell line and are a core component of the Expi293 Expression System.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A145271 mL
A145286 x 1 mL
Catalog number A14527
Price (USD)/ Each
1,174.65
Online exclusive
1,298.00 
Save 123.35 (10%)
-
Add to cart
Quantity:
1 mL
Price (USD)/ Each
1,174.65
Online exclusive
1,298.00 
Save 123.35 (10%)
Add to cart
Expi293F™ Cells
Catalog numberA14527
Price (USD)/ Each
1,174.65
Online exclusive
1,298.00 
Save 123.35 (10%)
-
Add to cart
Expi293F human cells are derived from the 293 cell line, and are a core component of the Expi293 Expression System. They are maintained in suspension culture and will grow to high density in Expi293 Expression Medium. Expi293F cells are highly transfectable and generate superior protein yields compared to standard 293 cell lines in transient protein expression.
Expi293F Cells attributes include:
Suspension culture cells pre-adapted to Expi293 Expression Medium
Demonstrated higher protein expression levels than other 293 cells adapted to Expi293 Expression Medium
Exhibit more rapid cell growth and higher culture viabilities than standard suspension-adapted 293 cells
Consistent transient expression performance of Expi293F cells for over 30 passages beyond thaw
• Also available from a cGMP bank (Cat. No. 100044202)
Frozen cells are supplied in a vial containing 1 ml of cells at 1 x 107 viable cells/ml in Expi293 Expression Medium and 10% DMSO. The cells should be thawed directly into Expi293 Expression Medium. The six-vial multipack eliminates the need to expand and rebank the cells in the lab.
Cells are typically grown in flasks on a shaker platform in a humidified, CO2 cell culture incubator. However, cultures are scalable from volumes of <1 mL in multiwell plates to >10 L in bioreactors. Protein harvest can be performed at 2 to 7 days post-transfection, depending on the protein expressed. For expression of recombinant antibodies, harvest is typically at 5 to 7 days post-transfection. There is no need for culture medium supplementation or batch feeding during expression.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell LineExpi293F
Cell TypeKidney (Embryonic)
Culture TypeSuspension Cell Culture
FormCryopreserved
SpeciesHuman
Media RecommendationExpi293™ Expression Medium
No. of Cells1 x 107
Product LineExpi293™
Product TypeExpi293F Cells
Quantity1 mL
Shipping ConditionDry Ice
SourceHuman
Unit SizeEach
Contents & Storage
1 x 107 cells⁄mL, store in liquid nitrogen