Thermo Scientific Chemicals

beta-Cyclodextrin hydrate, Thermo Scientific Chemicals

Catalog number: A14529.06
5 g, Each
Thermo Scientific Chemicals

beta-Cyclodextrin hydrate, Thermo Scientific Chemicals

Catalog number: A14529.06
5 g, Each
Quantity
Catalog number: A14529.06
also known as A14529-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
68168-23-0
IUPAC Name
5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol
Molecular Formula
C42H70O35
InChI Key
WHGYBXFWUBPSRW-UHFFFAOYSA-N
SMILES
OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Water Content (Karl Fischer Titration)
5.0-15.0%
Optical Rotation
+138.0 to +146.0° (c=1 in water)

Description

beta-Cyclodextrin hydrate forms clathrates. Produces a water-soluble complex with dansyl chloride for the fluorescent labeling of proteins. It is used in pharmaceutical research, as pharmaceutical intermediates and also as chemical reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
beta-Cyclodextrin hydrate forms clathrates. Produces a water-soluble complex with dansyl chloride for the fluorescent labeling of proteins. It is used in pharmaceutical research, as pharmaceutical intermediates and also as chemical reagents.

Solubility
Slightly soluble in water (18.5 g/L at 25°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text