Thermo Scientific Chemicals

2,4-Dinitrobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A14536.14
25 g, Each
Thermo Scientific Chemicals

2,4-Dinitrobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A14536.14
25 g, Each
Quantity
Catalog number: A14536.14
also known as A14536-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
610-30-0
IUPAC Name
2,4-dinitrobenzoic acid
Molecular Formula
C7H4N2O6
InChI Key
ZIIGSRYPZWDGBT-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O
Melting Point (clear melt)
179.0-185.0?C
Assay (Silylated GC)
≥97.5%
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

2,4-Dinitrobenzoic acid was used in the spectrophotometric determination of diazepam in pure samples and in its pharmaceutical preparations. It was employed as chromophore probe for analysis of perfluorinated carboxylic acids in water by capillary zone electrophoresis. It can be used to produce 2,4-dinitro-benzoyl chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dinitrobenzoic acid was used in the spectrophotometric determination of diazepam in pure samples and in its pharmaceutical preparations. It was employed as chromophore probe for analysis of perfluorinated carboxylic acids in water by capillary zone electrophoresis. It can be used to produce 2,4-dinitro-benzoyl chloride.

Solubility
Soluble in dioxane 50 mg/ml.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text