Thermo Scientific Chemicals

Fumaryl chloride, 95%, Thermo Scientific Chemicals

Catalog number: A14540.22
100 g, Each
Thermo Scientific Chemicals

Fumaryl chloride, 95%, Thermo Scientific Chemicals

Catalog number: A14540.22
100 g, Each
Quantity
Catalog number: A14540.22
also known as A14540-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
627-63-4
IUPAC Name
(2E)-but-2-enedioyl dichloride
Molecular Formula
C4H2Cl2O2
InChI Key
ZLYYJUJDFKGVKB-OWOJBTEDSA-N
SMILES
ClC(=O)C=CC(Cl)=O
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥94.0%
Refractive Index
1.4970-1.5030 @ 20°C
Form
Liquid
Identification (FTIR)
Conforms

Description

Fumaryl chloride was used in the preparation of high molecular weight poly(propylene fumarate). The photodissociation dynamics of fumaryl chloride was studied. It is used in agrochemicals and pharmaceuticals manufacturing. Also used as an intermediate for dyes, textile auxiliaries and peroxide compounds. It is an acid halide building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fumaryl chloride was used in the preparation of high molecular weight poly(propylene fumarate). The photodissociation dynamics of fumaryl chloride was studied. It is used in agrochemicals and pharmaceuticals manufacturing. Also used as an intermediate for dyes, textile auxiliaries and peroxide compounds. It is an acid halide building block.

Solubility
It reacts with water.

Notes
It is moisture sensitive. Incompatible with oxidizing agents, active metals and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text