2-Oxazolidinone is used as antimicrobials, in organisms it act as a protein synthesis inhibitors, targeting an early step involving the binding of N-formylmethionyl-tRNA to the ribosome. They are used for chiral synthesis. It has been used as an ethanolamine equivalent, also behaves as an aziridine equivalent as in the conversion of thiophenols or amines to their aminoethyl derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Oxazolidinone is used as antimicrobials, in organisms it act as a protein synthesis inhibitors, targeting an early step involving the binding of N-formylmethionyl-tRNA to the ribosome. They are used for chiral synthesis. It has been used as an ethanolamine equivalent, also behaves as an aziridine equivalent as in the conversion of thiophenols or amines to their aminoethyl derivatives.
Solubility
Soluble in water.
Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
Hideaki Kakeya; Masayuki Morishita; Hiroyuki Koshino; Tetsu-ichiro Morita; Kimiko Kobayashi and Hiroyuki Osada. Cytoxazone: A Novel Cytokine Modulator Containing a 2-Oxazolidinone Ring Produced by Streptomyces sp. J. Org. Chem. 1999, 64 (3), 1052-1053.
Guo-Jie Ho; David J. Mathre. Lithium-Initiated Imide Formation. A Simple Method for N-Acylation of 2-Oxazolidinones and Bornane-2,10-Sultam. J. Org. Chem. 1995, 60 (7), 2271-2273.
Has been used as an ethanolamine equivalent, e.g. in reaction with succinic anhydride to give 1-(2-hydroxyethyl)succinimide: Tetrahedron, 44, 261 (1988). Also behaves as an aziridine equivalent as in the conversion of thiophenols or amines to their aminoethyl derivatives: J. Org. Chem., 57, 6257 (1992).