Thermo Scientific Chemicals

Cadmium sulfide, 98%, Thermo Scientific Chemicals

Catalog number: A14544.0E
2500 g, Each
Thermo Scientific Chemicals

Cadmium sulfide, 98%, Thermo Scientific Chemicals

Catalog number: A14544.0E
2500 g, Each
Quantity
Catalog number: A14544.0E
also known as A14544-0E
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1306-23-6
IUPAC Name
cadmium(2+) sulfanediide
Molecular Formula
CdS
InChI Key
FRLJSGOEGLARCA-UHFFFAOYSA-N
SMILES
[S--].[Cd++]
Appearance (Color)
Orange
Form
Crystalline powder or powder
Assay from Supplier's CofA
≥97.5%
Water Content (Karl Fischer Titration)
<2% (KF from Supplier CofA)

Description

Cadmium sulfide is used as a pigment in the chemical industry; for manufacturing of photoresistors, optical-electronic pairs, photodiodes, solar batteries and phosphors. Single crystals of cadmium sulfide are used for the preparation of optical devices operating in the infrared regaion of spectrum. Cadmium sulfide is a direct band gap semiconductor with a band gap band from 2.42 to 2.57 eV.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cadmium sulfide is used as a pigment in the chemical industry; for manufacturing of photoresistors, optical-electronic pairs, photodiodes, solar batteries and phosphors. Single crystals of cadmium sulfide are used for the preparation of optical devices operating in the infrared regaion of spectrum. Cadmium sulfide is a direct band gap semiconductor with a band gap band from 2.42 to 2.57 eV.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and acids.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text