Cadmium sulfide, 98%, Thermo Scientific Chemicals
Cadmium sulfide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cadmium sulfide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A14544.22
also known as A14544-22
Price (USD)/ Each
39.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
46.10 
Save 6.45 (14%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
39.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
46.10 
Save 6.45 (14%)
Add to cart
Cadmium sulfide, 98%, Thermo Scientific Chemicals
Catalog numberA14544.22
Price (USD)/ Each
39.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
46.10 
Save 6.45 (14%)
-
Add to cart
Chemical Identifiers
CAS1306-23-6
IUPAC Namecadmium(2+) sulfanediide
Molecular FormulaCdS
InChI KeyFRLJSGOEGLARCA-UHFFFAOYSA-N
SMILES[S--].[Cd++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange
FormCrystalline powder or powder
Assay from Supplier's CofA≥97.5%
Water Content (Karl Fischer Titration)<2% (KF from Supplier CofA)
Cadmium sulfide is used as a pigment in the chemical industry; for manufacturing of photoresistors, optical-electronic pairs, photodiodes, solar batteries and phosphors. Single crystals of cadmium sulfide are used for the preparation of optical devices operating in the infrared regaion of spectrum. Cadmium sulfide is a direct band gap semiconductor with a band gap band from 2.42 to 2.57 eV.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cadmium sulfide is used as a pigment in the chemical industry; for manufacturing of photoresistors, optical-electronic pairs, photodiodes, solar batteries and phosphors. Single crystals of cadmium sulfide are used for the preparation of optical devices operating in the infrared regaion of spectrum. Cadmium sulfide is a direct band gap semiconductor with a band gap band from 2.42 to 2.57 eV.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and acids.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. N.Chestnoy; T.D.Harris; R.Hull; L.E.Brus. Luminescence and photophysics of cadmium sulfide semiconductor clusters: the nature of the emitting electronic state. J. Phys. Chem. 1986, 90, (15),3393-3399
  2. Norman Herron; Ying Wang; Hellmut Eckert. Synthesis and characterization of surface-capped, size-quantized cadmium sulfide clusters. Chemical control of cluster size. J. Am. Chem. Soc. 1990, 112, (4),1322-1326