Thermo Scientific Chemicals

p-Toluenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A14547.0B
1000 g, Each
Thermo Scientific Chemicals

p-Toluenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A14547.0B
1000 g, Each
Quantity
Catalog number: A14547.0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
98-59-9
IUPAC Name
4-methylbenzene-1-sulfonyl chloride
Molecular Formula
C7H7ClO2S
InChI Key
YYROPELSRYBVMQ-UHFFFAOYSA-N
SMILES
CC1=CC=C(C=C1)S(Cl)(=O)=O
Melting Point (clear melt)
64.0-72.0?C
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms

Description

p-Toluenesulfonyl chloride is used as a precursor in the production of dyes and saccharin. It acts as a dehydrating agent in the conversion of urea to carbodiimide and converts alcohols into the corresponding toluenesulfonate esters. It is also employed as a flow-promoting agent for paints, an adhesive, nitrocellulose, coating material and as a plasticizer for polyamides. Furthermore, it serves as an antistatic agent, as a gloss enhancer in plastic film preparations and as a basic material of electroplating solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Toluenesulfonyl chloride is used as a precursor in the production of dyes and saccharin. It acts as a dehydrating agent in the conversion of urea to carbodiimide and converts alcohols into the corresponding toluenesulfonate esters. It is also employed as a flow-promoting agent for paints, an adhesive, nitrocellulose, coating material and as a plasticizer for polyamides. Furthermore, it serves as an antistatic agent, as a gloss enhancer in plastic film preparations and as a basic material of electroplating solutions.

Solubility
Soluble in alcohol, benzene and ether. Insoluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text