Thermo Scientific Chemicals

Methyl carbazate, 98%, Thermo Scientific Chemicals

Catalog number: A14548.0E
2500 g, Each
Thermo Scientific Chemicals

Methyl carbazate, 98%, Thermo Scientific Chemicals

Catalog number: A14548.0E
2500 g, Each
Quantity
Catalog number: A14548.0E
also known as A14548-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6294-89-9
IUPAC Name
methoxycarbohydrazide
Molecular Formula
C2H6N2O2
InChI Key
WFJRIDQGVSJLLH-UHFFFAOYSA-N
SMILES
COC(=O)NN
Form
Crystals or platelets or powder or crystalline powder
Melting Point (clear melt)
68-73°
Appearance (Color)
White to pale cream to pink
Assay (GC)
≥97.5%

Description

Methyl carbazate was used in the preparation of methyl 3-(4-methyl-benzyl-idene)carbazate. It was also used in the synthesis of imidazo[1,5-d][1,2,4]triazines. It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl carbazate was used in the preparation of methyl 3-(4-methyl-benzyl-idene)carbazate. It was also used in the synthesis of imidazo[1,5-d][1,2,4]triazines. It is used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text