2-Methyl-3,5-dinitrobenzoic acid was used to synthesis heterocyclic derivatives and in the synthesis of isoquinolone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Methyl-3,5-dinitrobenzoic acid was used to synthesis heterocyclic derivatives and in the synthesis of isoquinolone.
Solubility
Soluble in water.
Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
Meth-Cohn O and Taljaard HC. Pyridine annelation of o-methyl arylcarboxylic acids with vilsmeier reagents. Tetrahedron Lett. 1983, 24 (42), 4607-4610.
Wu MT. J. Synthesis of some heterocyclic derivatives of 3, 5-dinitro o-toluic acid. Heterocycl. Chem. 1972, 9 (31), 693-705.