Thermo Scientific Chemicals

2-Methyl-3,5-dinitrobenzoic acid, 97+%, Thermo Scientific Chemicals

Catalog number: A14555.36
500 g, Each
Thermo Scientific Chemicals

2-Methyl-3,5-dinitrobenzoic acid, 97+%, Thermo Scientific Chemicals

Catalog number: A14555.36
500 g, Each
Quantity
Catalog number: A14555.36
also known as A14555-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
28169-46-2
Appearance (Color)
White to pale cream or pale yellow
Assay (Aqueous acid-base Titration)
≥97.0 to ≤103.0%
Melting Point (clear melt)
204-210°C
Form
Crystals or crystalline powder or powder
Assay (Silylated GC)
≥97.0%

Description

2-Methyl-3,5-dinitrobenzoic acid was used to synthesis heterocyclic derivatives and in the synthesis of isoquinolone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-3,5-dinitrobenzoic acid was used to synthesis heterocyclic derivatives and in the synthesis of isoquinolone.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text