2,6-Difluorobenzoic acid, 98+%, Thermo Scientific Chemicals
2,6-Difluorobenzoic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,6-Difluorobenzoic acid, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A14558.14
also known as A14558-14
Price (USD)/ Each
104.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
104.00
Add to cart
2,6-Difluorobenzoic acid, 98+%, Thermo Scientific Chemicals
Catalog numberA14558.14
Price (USD)/ Each
104.00
-
Add to cart
Chemical Identifiers
CAS385-00-2
IUPAC Name2,6-difluorobenzoic acid
Molecular FormulaC7H4F2O2
InChI KeyONOTYLMNTZNAQZ-UHFFFAOYSA-N
SMILESOC(=O)C1=C(F)C=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)155.5-161.5?C
Appearance (Color)White to cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only