2-Naphthol can be used as a fluorescent indicator. It can be also used in the production of dyes and in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Naphthol can be used as a fluorescent indicator. It can be also used in the production of dyes and in organic synthesis.
Solubility
Partly miscible in water (0.76 g/L).
Notes
It is sensitive to light. Incompatible with oxidizing agents.
RUO – Research Use Only
M. Panizza; P.A.Michaud, G.Cerisola; Ch. Comninellis. Anodic oxidation of 2-naphthol at boron-doped diamond electrodes. Journal of Electroanalytical Chemistry. 2001, 507, (1-2),206-214
William R.Laws; Ludwig. Brand. Analysis of two-state excited-state reactions. The fluorescence decay of 2-naphthol. J. Phys. Chem. 1979, 83, (7),795-802