Thermo Scientific Chemicals

3-(4-Hydroxyphenyl)propionic acid, 99%, Thermo Scientific Chemicals

Catalog number: A14567.09
10 g, Each
Thermo Scientific Chemicals

3-(4-Hydroxyphenyl)propionic acid, 99%, Thermo Scientific Chemicals

Catalog number: A14567.09
10 g, Each
Quantity
Catalog number: A14567.09
also known as A14567-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
501-97-3
IUPAC Name
3-(4-hydroxyphenyl)propanoic acid
Molecular Formula
C9H10O3
InChI Key
NMHMNPHRMNGLLB-UHFFFAOYSA-N
SMILES
OC(=O)CCC1=CC=C(O)C=C1
Form
Crystalline powder or powder or granules
Melting Point (clear melt)
126-133?C
Appearance (Color)
White to cream to pale brown
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Identification (FTIR)
Conforms

Description

3-(4-Hydroxyphenyl)propionic acid is a potential antioxidant. It is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(4-Hydroxyphenyl)propionic acid is a potential antioxidant. It is used as pharmaceutical intermediates.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text