Thermo Scientific Chemicals

n-Tricosane, 99%, Thermo Scientific Chemicals

Catalog number: A14577.14
25 g, Each
Thermo Scientific Chemicals

n-Tricosane, 99%, Thermo Scientific Chemicals

Catalog number: A14577.14
25 g, Each
Quantity
Catalog number: A14577.14
also known as A14577-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
638-67-5
IUPAC Name
tricosane
Molecular Formula
C23H48
InChI Key
FIGVVZUWCLSUEI-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCCCCCCC
Melting Point (clear melt)
45.0-51.0?C
Appearance (Color)
White
Assay (GC)
≥98.5%
Form
Crystals or powder or crystalline powder or flakes or waxy solid
Identification (FTIR)
Conforms

Description

n-Tricosane is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Tricosane is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text