Thermo Scientific Chemicals

2'-Hydroxy-4',6'-dimethoxyacetophenone, 98%, Thermo Scientific Chemicals

Catalog number: A14585.03
1 g, Each
Thermo Scientific Chemicals

2'-Hydroxy-4',6'-dimethoxyacetophenone, 98%, Thermo Scientific Chemicals

Catalog number: A14585.03
1 g, Each
Quantity
Catalog number: A14585.03
also known as A14585-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
90-24-4
IUPAC Name
1-(2-hydroxy-4,6-dimethoxyphenyl)ethan-1-one
Molecular Formula
C10H12O4
InChI Key
FBUBVLUPUDBFME-UHFFFAOYSA-N
SMILES
COC1=CC(O)=C(C(C)=O)C(OC)=C1
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Appearance (Color)
White to cream
Melting Point
79.5-83.5?C

Description

2'-Hydroxy-4',6'-dimethoxyacetophenone is a chemical with potential antibacterial agent effects and urease inhibitory effects as well as synthesis applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Hydroxy-4′,6′-dimethoxyacetophenone is a chemical with potential antibacterial agent effects and urease inhibitory effects as well as synthesis applications.

Solubility
Soluble in hot methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text