Thermo Scientific Chemicals

4-Hydroxy-2-methylquinoline, 98+%, Thermo Scientific Chemicals

Catalog number: A14587.06
5 g, Each
Thermo Scientific Chemicals

4-Hydroxy-2-methylquinoline, 98+%, Thermo Scientific Chemicals

Catalog number: A14587.06
5 g, Each
Quantity
Catalog number: A14587.06
also known as A14587-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
607-67-0
IUPAC Name
2-methyl-1,4-dihydroquinolin-4-one
Molecular Formula
C10H9NO
InChI Key
NWINIEGDLHHNLH-UHFFFAOYSA-N
SMILES
CC1=CC(=O)C2=CC=CC=C2N1
Appearance (Color)
Pale cream to cream to pale brown
Form
Powder
Melting Point (clear melt)
230.5-236.5?C
Assay (HPLC)
≥98.0%

Description

4-Hydroxy-2-methylquinoline is act as an intermediate in the synthesis of dequalinium chloride and as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxy-2-methylquinoline is act as an intermediate in the synthesis of dequalinium chloride and as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text