Zinc carbonate basic, 97%, Zn >58.0%, Thermo Scientific Chemicals
Zinc carbonate basic, 97%, Zn >58.0%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc carbonate basic, 97%, Zn >58.0%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
250 g
Catalog number A14590.0B
also known as A14590-0B
Price (USD)/ Each
101.65
Online exclusive
113.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (USD)/ Each
101.65
Online exclusive
113.00 
Save 11.35 (10%)
Add to cart
Zinc carbonate basic, 97%, Zn >58.0%, Thermo Scientific Chemicals
Catalog numberA14590.0B
Price (USD)/ Each
101.65
Online exclusive
113.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS5263-02-5
IUPAC Namepentazinc(2+) hexahydroxide dicarbonate
Molecular FormulaC2H6O12Zn5
InChI KeyUOURRHZRLGCVDA-UHFFFAOYSA-D
SMILES[OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Zn++].[Zn++].[Zn++].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Formpowder
Complexometric Titration> 58.0% Zn (dry wt. basis)
Loss on Drying< 3.0% (105?C, 2hrs)
Zinc carbonate basic is used as pharmaceutical intermediate and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc carbonate basic is used as pharmaceutical intermediate and in chemical research.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Yanhua Liu; Jingzhe Zhao; Hui Zhang; Yanchao Zhu; Zichen Wang. Thermal decomposition of basic zinc carbonate in nitrogen atmosphere. Thermochimica Acta. 2004, 414, (2),121-123
  2. D.Klissurski; I.Uzunov; K.Kumbilieva. Preparation of highly dispersed zinc oxide by thermal decomposition of basic zinc carbonate. Thermochimica Acta. 1985, 93, 485-488