Thermo Scientific Chemicals

Zinc carbonate basic, 97%, Zn >58.0%, Thermo Scientific Chemicals

Catalog number: A14590.30
250 g, Each
Thermo Scientific Chemicals

Zinc carbonate basic, 97%, Zn >58.0%, Thermo Scientific Chemicals

Catalog number: A14590.30
250 g, Each
Quantity
Catalog number: A14590.30
also known as A14590-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5263-02-5
IUPAC Name
pentazinc(2+) hexahydroxide dicarbonate
Molecular Formula
C2H6O12Zn5
InChI Key
UOURRHZRLGCVDA-UHFFFAOYSA-D
SMILES
[OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Zn++].[Zn++].[Zn++].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O
Appearance (Color)
White
Form
powder
Complexometric Titration
> 58.0% Zn (dry wt. basis)
Loss on Drying
< 3.0% (105?C, 2hrs)

Description

Zinc carbonate basic is used as pharmaceutical intermediate and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc carbonate basic is used as pharmaceutical intermediate and in chemical research.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text