Thermo Scientific Chemicals

Sarcosine, 98%

Catalog number: A14594.22
100 g, Each
Thermo Scientific Chemicals

Sarcosine, 98%

Catalog number: A14594.22
100 g, Each
Quantity
Catalog number: A14594.22
also known as A14594-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
107-97-1
Appearance (Color)
White
Identification (FTIR)
Conforms
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Water Content (Karl Fischer Titration)
≤1%
Form
Crystals or powder or crystalline powder or fused solid

Description

A type 1 glycine transporter inhibitorSarcosine is used in the production of biodegradable surfactants and toothpastes. It is used as an intermediate in the synthesis of antienzyme agents. It is found to be a marker for prostate cancer bioagression. It acts as a biochemical reagent, dye stabilizer, daily chemical, amino acid surfactants, health drugs fatigue recovery agent and so on. It is involved in the preparation of N-methyl-glycine ethyl ester by reacting with ethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A type 1 glycine transporter inhibitorSarcosine is used in the production of biodegradable surfactants and toothpastes. It is used as an intermediate in the synthesis of antienzyme agents. It is found to be a marker for prostate cancer bioagression. It acts as a biochemical reagent, dye stabilizer, daily chemical, amino acid surfactants, health drugs fatigue recovery agent and so on. It is involved in the preparation of N-methyl-glycine ethyl ester by reacting with ethanol.

Solubility
Soluble in water, methanol. Slightly soluble in ether.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text