Thermo Scientific Chemicals

Maleic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A14596.0I
5000 g, Each
Thermo Scientific Chemicals

Maleic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A14596.0I
5000 g, Each
Quantity
Catalog number: A14596.0I
also known as A14596-0I
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-16-7
IUPAC Name
(2E)-but-2-enedioic acid
Molecular Formula
C4H4O4
InChI Key
VZCYOOQTPOCHFL-OWOJBTEDSA-N
SMILES
OC(=O)\C=C\C(O)=O
Form
Crystals or powder or crystalline powder
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
130-138?C

Description

Maleic acid is used as a precursor to fumaric acid, dimethyl maleate and glyoxalic acid. It is an electrophile and acts as dienophine in Diels-Alder reactions. It reacts with drugs to form more stable addition salts like indacaterol maleate, carfenazine, chlorpheniramine, pyrilamine, methylergonovine and thiethylperazine. Its maleate ion is useful in biochemistry as an inhibitor of transaminase reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Maleic acid is used as a precursor to fumaric acid, dimethyl maleate and glyoxalic acid. It is an electrophile and acts as dienophine in Diels-Alder reactions. It reacts with drugs to form more stable addition salts like indacaterol maleate, carfenazine, chlorpheniramine, pyrilamine, methylergonovine and thiethylperazine. Its maleate ion is useful in biochemistry as an inhibitor of transaminase reactions.

Solubility
Soluble in water, acetone, glacial acetic acid, alcohol and concentrated sulfuric acid. Slightly soluble in diethyl ether. Insoluble in benzene and chloroform.

Notes
Incompatible with oxidizing agents, bases, reducing agents, metals and alkalis.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text